Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AHL19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111328.3 | NP_566232.1 | 1 | 1381 | 98 | |
GenBank mRNA | AK228206.1 | - | 1 | 1141 | 98 | |
AY085572.1 | - | 1 | 1133 | 98 | ||
BT005882.1 | - | 1 | 906 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |