Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111406.5 | NP_187184.2 | 3 - 4 | 1040 | 151 | |
GenBank mRNA | AK175941.1 | - | 3 - 4 | 625 | 151 | |
AK227452.1 | - | 3 - 4 | 586 | 151 | ||
BT006478.1 | - | 3 - 4 | 290 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |