Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202886.1 | NP_001189815.1 | 8 - 9 | 1143 | 75 | |
NM_001337585.1 | NP_001319478.1 | 7 - 8 | 1122 | 75 | ||
GenBank mRNA | BT029213.1 | - | 7 - 8 | 1011 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |