Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMPA-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337604.1 | NP_001319482.1 | 6 - 7 | 935 | 61 | |
NM_111446.2 | NP_187223.1 | 6 - 7 | 870 | 61 | ||
GenBank mRNA | DQ446636.1 | - | 6 - 7 | 850 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |