Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRXIIF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337629.1 | NP_001327152.1 | 2 - 3 | 357 | 90 | |
NM_111480.4 | NP_566268.1 | 2 - 3 | 346 | 90 | ||
GenBank mRNA | AY052278.1 | - | 2 - 3 | 279 | 90 | |
AY098963.1 | - | 2 - 3 | 249 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |