Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111566.4 | NP_187342.1 | 9 - 10 | 1092 | 63 | |
GenBank mRNA | AF123254.1 | - | 9 - 10 | 901 | 63 | |
AY062621.1 | - | 9 - 10 | 958 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |