Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCPL27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111678.3 | NP_187456.1 | 4 - 5 | 793 | 102 | |
GenBank mRNA | BT015083.1 | - | 4 - 5 | 702 | 102 | |
BT015904.1 | - | 4 - 5 | 643 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |