Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATAB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111680.4 | NP_566327.1 | 1 - 2 | 557 | 131 | |
GenBank mRNA | AY072316.1 | - | 1 - 2 | 499 | 131 | |
BT001168.1 | - | 1 - 2 | 481 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |