Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337758.1 | NP_001319499.1 | 8 - 9 | 1331 | 114 | |
NM_111683.2 | NP_187461.1 | 7 - 8 | 1416 | 114 | ||
GenBank mRNA | AF448231.1 | - | 8 - 9 | 1230 | 114 | |
AY056439.1 | - | 7 - 8 | 1215 | 114 | ||
AY057517.1 | - | 7 - 8 | 1215 | 114 | ||
AY133652.1 | - | 7 - 8 | 1113 | 114 | ||
BT006370.1 | - | 7 - 8 | 1113 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |