Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSLG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341637.1 | NP_001328486.1 | 1 - 2 | 258 | 110 | |
NM_118532.4 | NP_567692.2 | 2 - 3 | 958 | 110 | ||
GenBank mRNA | AY070072.1 | - | 2 - 3 | 811 | 110 | |
AY096446.1 | - | 2 - 3 | 723 | 110 | ||
KJ138739.1 | - | 2 - 3 | 723 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |