Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118557.4 | NP_194155.1 | 2 - 3 | 1289 | 128 | |
GenBank mRNA | AF272746.1 | - | 2 - 3 | 997 | 128 | |
AY072151.1 | - | 2 - 3 | 1061 | 128 | ||
AY084583.1 | - | 2 - 3 | 1059 | 128 | ||
AY096490.1 | - | 2 - 3 | 897 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |