Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341743.1 | NP_001328956.1 | 2 - 3 | 766 | 77 | |
NM_001341744.1 | NP_001328957.1 | 3 - 4 | 672 | 77 | ||
NM_118676.4 | NP_194274.2 | 3 - 4 | 665 | 77 | ||
GenBank mRNA | AJ252063.1 | - | 2 - 3 | 529 | 77 | |
BT026122.1 | - | 2 - 3 | 529 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |