Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036637.2 | NP_001031714.1 | 9 - 10 | 1197 | 102 | |
NM_001084977.1 | NP_001078446.1 | 9 - 10 | 1190 | 102 | ||
NM_118677.5 | NP_194275.2 | 9 - 10 | 1311 | 102 | ||
GenBank mRNA | AK175486.1 | - | 9 - 10 | 1195 | 102 | |
AY094470.1 | - | 9 - 10 | 1192 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |