Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACHT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118749.4 | NP_194346.1 | 2 - 3 | 449 | 109 | |
GenBank mRNA | BT024608.1 | - | 2 - 3 | 404 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |