Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIOX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203915.2 | NP_001190844.1 | 4 - 5 | 592 | 97 | |
NM_118759.5 | NP_194356.2 | 4 - 5 | 697 | 97 | ||
GenBank mRNA | AY142501.1 | - | 4 - 5 | 453 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |