Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FIM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036649.1 | NP_001031726.1 | 12 - 13 | 1938 | 70 | |
NM_001036650.2 | NP_001031727.1 | 11 - 12 | 2025 | 70 | ||
NM_001341826.1 | NP_001328753.1 | 12 - 13 | 2065 | 70 | ||
NM_001341827.1 | NP_001328754.1 | 12 - 13 | 2049 | 70 | ||
NM_001341828.1 | NP_001328752.1 | 12 - 13 | 2037 | 70 | ||
NM_001341829.1 | NP_001328755.1 | 10 - 11 | 1959 | 70 | ||
NM_118804.5 | NP_194400.1 | 11 - 12 | 1889 | 70 | ||
GenBank mRNA | AK318682.1 | - | 12 - 13 | 1941 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |