Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FIM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036649.1 | NP_001031726.1 | 5 - 6 | 992 | 126 | |
NM_001036650.2 | NP_001031727.1 | 4 - 5 | 1079 | 126 | ||
NM_001341826.1 | NP_001328753.1 | 5 - 6 | 1119 | 126 | ||
NM_001341827.1 | NP_001328754.1 | 5 - 6 | 1103 | 126 | ||
NM_001341828.1 | NP_001328752.1 | 5 - 6 | 1091 | 126 | ||
NM_001341829.1 | NP_001328755.1 | 3 - 4 | 1013 | 126 | ||
NM_118804.5 | NP_194400.1 | 4 - 5 | 943 | 126 | ||
GenBank mRNA | AK318682.1 | - | 5 - 6 | 995 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |