Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125593.1 | NP_001119065.1 | 3 - 4 | 739 | 87 | |
NM_001125594.1 | NP_001119066.1 | 3 - 4 | 552 | 87 | ||
NM_001125595.2 | NP_001119067.1 | 5 - 6 | 655 | 87 | ||
NM_001341834.1 | NP_001328604.1 | 3 - 4 | 917 | 87 | ||
NM_001341835.1 | NP_001328603.1 | 3 - 4 | 897 | 87 | ||
NM_118838.5 | NP_194434.2 | 5 - 6 | 706 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |