Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VIIIB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341848.1 | NP_001328308.1 | 13 - 14 | 1688 | 85 | |
NM_001341849.1 | NP_001328309.1 | 12 - 13 | 1627 | 85 | ||
NM_001341850.1 | NP_001328307.1 | 12 - 13 | 1704 | 85 | ||
NM_118871.6 | NP_194467.5 | 11 - 12 | 1495 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |