Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148863.4 | NP_680116.2 | 1 - 2 | 1236 | 104 | |
GenBank mRNA | AY664499.1 | - | 1 - 2 | 883 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |