Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBP13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202935.1 | NP_001189864.1 | 2 - 3 | 365 | 81 | |
NM_001337961.1 | NP_001319526.1 | 2 - 3 | 365 | 81 | ||
NM_001337962.1 | NP_001326292.1 | 2 - 3 | 368 | 81 | ||
NM_112024.6 | NP_187797.3 | 2 - 3 | 294 | 81 | ||
GenBank mRNA | BT002760.1 | - | 2 - 3 | 95 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |