Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202939.1 | NP_001189868.1 | 12 - 13 | 2508 | 119 | |
NM_112064.5 | NP_566417.3 | 12 - 13 | 2697 | 119 | ||
GenBank mRNA | AF245395.1 | - | 12 - 13 | 2415 | 119 | |
AY150438.1 | - | 11 - 12 | 2211 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |