Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ckl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119030.4 | NP_194615.2 | 12 - 13 | 1204 | 70 | |
GenBank mRNA | AK118912.1 | - | 12 - 13 | 1125 | 70 | |
AK175508.1 | - | 12 - 13 | 1173 | 70 | ||
AK176345.1 | - | 12 - 13 | 1167 | 70 | ||
AK176894.1 | - | 12 - 13 | 1173 | 70 | ||
AY099731.1 | - | 12 - 13 | 1178 | 70 | ||
AY943853.1 | - | 12 - 13 | 1105 | 70 | ||
BT002172.1 | - | 12 - 13 | 1105 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |