Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HXK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119057.4 | NP_194642.1 | 6 - 7 | 1405 | 80 | |
GenBank mRNA | AK230265.1 | - | 3232 | 80 | ||
AY075658.1 | - | 6 - 7 | 1295 | 80 | ||
AY124809.1 | - | 6 - 7 | 1069 | 80 | ||
U28214.1 | - | 6 - 7 | 1266 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |