Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341965.1 | NP_001328427.1 | 3 - 4 | 501 | 110 | |
NM_119078.5 | NP_194662.2 | 3 - 4 | 609 | 110 | ||
GenBank mRNA | AY063817.1 | - | 3 - 4 | 342 | 110 | |
AY096457.1 | - | 3 - 4 | 225 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |