Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRE11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345081.1 | NP_001330550.1 | 7 - 8 | 795 | 62 | |
NM_124806.2 | NP_200237.1 | 9 - 10 | 981 | 62 | ||
GenBank mRNA | AJ243822.1 | - | 9 - 10 | 950 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |