Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124958.3 | NP_200387.1 | 4 - 5 | 403 | 143 | |
GenBank mRNA | AF283757.1 | - | 4 - 5 | 363 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |