Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036636.2 | NP_001031713.1 | 1 - 2 | 327 | 67 | |
NM_001341732.1 | NP_001329739.1 | 1 - 2 | 291 | 67 | ||
NM_001341733.1 | NP_001329738.1 | 1 - 2 | 263 | 67 | ||
NM_001341734.1 | NP_001329741.1 | 1 - 2 | 263 | 67 | ||
NM_001341738.1 | NP_001329742.1 | 1 - 2 | 291 | 67 | ||
NM_001341739.1 | NP_001329745.1 | 1 - 2 | 291 | 67 | ||
NM_001341740.1 | NP_001329744.1 | 1 - 2 | 291 | 67 | ||
NM_001341741.1 | NP_001329748.1 | 1 - 2 | 291 | 67 | ||
NM_001341742.1 | NP_001329747.1 | 1 - 2 | 291 | 67 | ||
NM_179112.3 | NP_849443.2 | 1 - 2 | 291 | 67 | ||
GenBank mRNA | BT010765.1 | - | 1 - 2 | 255 | 67 | |
BT010987.1 | - | 1 - 2 | 163 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |