Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLDDELTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119745.3 | NP_567989.1 | 3 - 4 | 1585 | 97 | |
NM_179170.3 | NP_849501.1 | 3 - 4 | 1789 | 97 | ||
NM_179171.2 | NP_849502.1 | 3 - 4 | 1552 | 97 | ||
GenBank mRNA | AB031047.1 | - | 3 - 4 | 1399 | 97 | |
AF306345.1 | - | 3 - 4 | 1263 | 97 | ||
AF322228.1 | - | 3 - 4 | 1272 | 97 | ||
AK227213.1 | - | 3 - 4 | 1395 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |