Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123759.1 | NP_001117231.1 | 4 - 5 | 659 | 71 | |
NM_001197982.1 | NP_001184911.1 | 6 - 7 | 954 | 71 | ||
NM_001331528.1 | NP_001322680.1 | 5 - 6 | 944 | 71 | ||
NM_001331529.1 | NP_001322678.1 | 4 - 5 | 811 | 71 | ||
NM_001331530.1 | NP_001322679.1 | 5 - 6 | 857 | 71 | ||
NM_001331531.1 | NP_001322673.1 | 4 - 5 | 2031 | 71 | ||
NM_001331532.1 | NP_001322674.1 | 5 - 6 | 1940 | 71 | ||
NM_001331533.1 | NP_001322675.1 | 5 - 6 | 1913 | 71 | ||
NM_001331534.1 | NP_001322676.1 | 3 - 4 | 1618 | 71 | ||
NM_001331535.1 | NP_001322677.1 | 4 - 5 | 1527 | 71 | ||
NM_100402.3 | NP_172015.1 | 4 - 5 | 1500 | 71 | ||
NM_179265.2 | NP_849596.1 | 5 - 6 | 963 | 71 | ||
GenBank mRNA | AK316776.1 | - | 5 - 6 | 868 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |