Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HUA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112103.3 | NP_187874.2 | 7 - 8 | 1204 | 87 | |
GenBank mRNA | AY024357.1 | - | 7 - 8 | 1204 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |