Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035611.2 | NP_001030688.1 | 3 - 4 | 431 | 73 | |
NM_112190.5 | NP_187953.1 | 3 - 4 | 434 | 73 | ||
GenBank mRNA | AB493611.1 | - | 2 - 3 | 169 | 73 | |
AF324696.3 | - | 3 - 4 | 433 | 73 | ||
AF327429.1 | - | 3 - 4 | 429 | 73 | ||
AF349523.1 | - | 2 - 3 | 169 | 73 | ||
AY054608.1 | - | 3 - 4 | 429 | 73 | ||
AY072455.1 | - | 2 - 3 | 169 | 73 | ||
AY084881.1 | - | 3 - 4 | 419 | 73 | ||
AY463625.1 | - | 2 - 3 | 169 | 73 | ||
X54996.1 | - | 396 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |