Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OVA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112195.3 | NP_187958.1 | 2 - 3 | 537 | 136 | |
GenBank mRNA | BT002317.1 | - | 2 - 3 | 489 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |