Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OVA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112195.3 | NP_187958.1 | 3 - 4 | 648 | 91 | |
GenBank mRNA | BT002317.1 | - | 3 - 4 | 600 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |