Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSP60-3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112240.4 | NP_566466.1 | 5 - 6 | 735 | 67 | |
GenBank mRNA | AY094453.1 | - | 5 - 6 | 649 | 67 | |
BT015923.1 | - | 5 - 6 | 533 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |