Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112293.5 | NP_188052.2 | 7 - 8 | 813 | 74 | |
NM_202582.1 | NP_974311.1 | 6 - 7 | 869 | 74 | ||
NM_202583.1 | NP_974312.1 | 7 - 8 | 737 | 74 | ||
GenBank mRNA | AY518292.1 | - | 7 - 8 | 807 | 74 | |
BT046202.1 | - | 7 - 8 | 569 | 74 | ||
FJ708721.1 | - | 7 - 8 | 569 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |