Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WAG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112295.4 | NP_188054.1 | 1 | 1371 | 121 | |
GenBank mRNA | AY088832.1 | - | 1 | 1263 | 121 | |
BT029242.1 | - | 1 | 1227 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |