Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSL04 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202964.2 | NP_001189893.1 | 12 - 13 | 1563 | 84 | |
NM_001338124.1 | NP_001326811.1 | 12 - 13 | 1683 | 84 | ||
NM_112317.3 | NP_188075.2 | 12 - 13 | 1722 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |