Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202966.1 | NP_001189895.1 | 5 - 6 | 2561 | 90 | |
NM_112342.4 | NP_188099.2 | 4 - 5 | 2653 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |