Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBP18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119316.5 | NP_194895.1 | 4 - 5 | 720 | 57 | |
GenBank mRNA | AY142532.1 | - | 4 - 5 | 613 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |