Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CESA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119393.3 | NP_194967.1 | 11 - 12 | 2561 | 102 | |
GenBank mRNA | AK226243.1 | - | 11 - 12 | 2240 | 102 | |
BT008654.1 | - | 11 - 12 | 2249 | 102 | ||
KJ138727.1 | - | 11 - 12 | 2105 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |