Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AME3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036697.1 | NP_001031774.1 | 6 - 7 | 580 | 121 | |
NM_001342166.1 | NP_001328058.1 | 4 - 5 | 908 | 121 | ||
NM_119418.3 | NP_194992.1 | 5 - 6 | 816 | 121 | ||
NM_202937.1 | NP_974666.1 | 5 - 6 | 706 | 121 | ||
GenBank mRNA | BT029776.1 | - | 5 - 6 | 438 | 121 | |
D45355.1 | - | 5 - 6 | 497 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |