Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDA14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119501.4 | NP_567921.1 | 6 - 7 | 651 | 67 | |
GenBank mRNA | AY052234.1 | - | 6 - 7 | 605 | 67 | |
AY113069.1 | - | 6 - 7 | 540 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |