Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DML3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125640.1 | NP_001119112.1 | 2 - 3 | 190 | 74 | |
NM_001342250.1 | NP_001329149.1 | 3 - 4 | 417 | 74 | ||
NM_001342252.1 | NP_001329148.1 | 1 - 2 | 290 | 74 | ||
NM_119567.4 | NP_195132.3 | 2 - 3 | 344 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |