Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XDH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342310.1 | NP_001329029.1 | 3 - 4 | 1774 | 85 | |
NM_001342311.1 | NP_001329028.1 | 4 - 5 | 1843 | 85 | ||
NM_119656.3 | NP_195216.2 | 4 - 5 | 1874 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |