Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179684.4 | NP_850015.1 | 2 - 3 | 533 | 61 | |
NM_179685.2 | NP_850016.1 | 2 - 3 | 503 | 61 | ||
GenBank mRNA | AB039925.1 | - | 2 - 3 | 364 | 61 | |
AY087097.1 | - | 2 - 3 | 485 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |