Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAT12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084501.1 | NP_001077970.1 | 1 - 2 | 802 | 147 | |
NM_001336128.1 | NP_001323574.1 | 1 - 2 | 802 | 147 | ||
NM_179777.2 | NP_850108.1 | 1 - 2 | 804 | 147 | ||
NM_201821.2 | NP_973550.2 | 1 - 2 | 802 | 147 | ||
GenBank mRNA | AY099545.1 | - | 1 - 2 | 715 | 147 | |
AY444865.1 | - | 1 - 2 | 544 | 147 | ||
BT002121.1 | - | 1 - 2 | 544 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |