Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THIC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202705.1 | NP_001189634.1 | 4 - 5 | 1443 | 86 | |
NM_001336219.1 | NP_001325132.1 | 1 - 2 | 905 | 86 | ||
NM_128517.4 | NP_180524.1 | 4 - 5 | 1399 | 86 | ||
NM_179804.3 | NP_850135.1 | 4 - 5 | 1399 | 86 | ||
GenBank mRNA | AY092989.1 | - | 4 - 5 | 1344 | 86 | |
AY128756.1 | - | 3 - 4 | 1252 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |