Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UVR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035626.2 | NP_001030703.1 | 8 - 9 | 1125 | 69 | |
NM_112432.2 | NP_566520.1 | 8 - 9 | 1300 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |