Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRZ4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338223.1 | NP_001327485.1 | 1 - 2 | 448 | 73 | |
NM_112497.3 | NP_188247.2 | 1 - 2 | 448 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |